Assay Details
| Gene Symbol: LOC100345975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721255.3 | XP_002721301.2 | 1381 | 69 | ||
| XM_017350778.1 | XP_017206267.1 | 1400 | 69 | |||
More Information
Set Membership: |
|

LOC100345975
Genomic Map