Assay Details
| Gene Symbol: MRPS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271255.2 | XP_008269477.2 | 1 - 2 | 134 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPS23| Gene Symbol: MRPS23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271255.2 | XP_008269477.2 | 1 - 2 | 134 | 60 | |
Set Membership: |
|