Assay Details
| Gene Symbol: RDH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254762.2 | XP_008252984.2 | 1 - 2 | 468 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RDH14| Gene Symbol: RDH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254762.2 | XP_008252984.2 | 1 - 2 | 468 | 76 | |
Set Membership: |
|