Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261251.2 | XP_008259473.1 | 3 - 4 | 1125 | 73 | |
| XM_017344139.1 | XP_017199628.1 | 3 - 4 | 872 | 73 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map