Assay Details
| Gene Symbol: LOC100357845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263811.2 | XP_008262033.1 | 1 - 2 | 108 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008263811.2 transcripts.
Set Membership: |
|

LOC100357845
Genomic Map