Assay Details
| Gene Symbol: WTAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263747.2 | XP_008261969.1 | 6 | 584 | 70 | |
| XM_017345471.1 | XP_017200960.1 | 6 | 583 | 70 | ||
More Information
Set Membership: |
|

WTAP
Genomic Map