Assay Details
| Gene Symbol: TTC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713995.3 | XP_002714041.1 | 2 - 3 | 340 | 66 | |
| XM_008262146.2 | XP_008260368.1 | 2 - 3 | 325 | 66 | ||
| XM_008262147.2 | XP_008260369.1 | 2 - 3 | 248 | 66 | ||
| XM_008262149.2 | XP_008260371.1 | 2 - 3 | 325 | 66 | ||
More Information
Set Membership: |
|

TTC33
Genomic Map