Assay Details
| Gene Symbol: LOC100338745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262321.2 | XP_008260543.2 | 10 - 11 | 776 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100338745| Gene Symbol: LOC100338745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262321.2 | XP_008260543.2 | 10 - 11 | 776 | 77 | |
Set Membership: |
|