Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719692.3 | XP_002719738.1 | 3 - 4 | 602 | 66 | |
| XM_008272133.2 | XP_008270355.1 | 3 - 4 | 549 | 66 | ||
More Information
Set Membership: |
|

MTFP1
Genomic Map