Assay Details
| Gene Symbol: SHOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266391.2 | XP_008264613.1 | 3 - 4 | 456 | 82 | |
| GenBank mRNA | KP726285.1 | - | 3 - 4 | 406 | 82 | |
More Information
Set Membership: |
|

SHOX2
Genomic Map