Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261531.2 | XP_008259753.1 | 2 - 3 | 382 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008261531.2 transcripts.
Set Membership: |
|

CCDC126
Genomic Map