Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261531.2 | XP_008259753.1 | 3 - 4 | 762 | 66 | |
| XM_008261533.2 | XP_008259755.1 | 4 - 5 | 915 | 66 | ||
| XM_008261534.2 | XP_008259756.1 | 2 - 3 | 1396 | 66 | ||
| XM_008261535.2 | XP_008259757.1 | 2 - 3 | 644 | 66 | ||
More Information
Set Membership: |
|

CCDC126
Genomic Map