Assay Details
| Gene Symbol: DDX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723122.3 | XP_002723168.1 | 1463 | 62 | ||
| XM_017339135.1 | XP_017194624.1 | 1464 | 62 | |||
| XM_017339136.1 | XP_017194625.1 | 1465 | 62 | |||
More Information
Set Membership: |
|

DDX24
Genomic Map