Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715775.3 | XP_002715821.1 | 7 - 8 | 632 | 71 | |
| XM_017345982.1 | XP_017201471.1 | 7 - 8 | 660 | 71 | ||
| XM_017345983.1 | XP_017201472.1 | 7 - 8 | 631 | 71 | ||
More Information
Set Membership: |
|

RAP1A
Genomic Map