Assay Details
| Gene Symbol: LIPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718369.3 | XP_002718415.2 | 6 - 7 | 1222 | 62 | |
| XM_008270162.2 | XP_008268384.1 | 8 - 9 | 1018 | 62 | ||
| XM_008270163.2 | XP_008268385.1 | 7 - 8 | 903 | 62 | ||
| XM_017348797.1 | XP_017204286.1 | 7 - 8 | 979 | 62 | ||
| XM_017348798.1 | XP_017204287.1 | 4 - 5 | 911 | 62 | ||
More Information
Set Membership: |
|

LIPM
Genomic Map