Assay Details
| Gene Symbol: LOC100351762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264430.2 | XP_008262652.1 | 2 - 3 | 210 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100351762| Gene Symbol: LOC100351762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264430.2 | XP_008262652.1 | 2 - 3 | 210 | 75 | |
Set Membership: |
|