Assay Details
| Gene Symbol: RPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275206.2 | XP_008273428.1 | 1 | 122 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008275206.2 transcripts.
Set Membership: |
|

RPL21
Genomic Map