Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719265.3 | XP_002719311.1 | 7 - 8 | 1953 | 65 | |
| XM_017349082.1 | XP_017204571.1 | 7 - 8 | 1953 | 65 | ||
More Information
Set Membership: |
|

RNF43
Genomic Map