Assay Details
| Gene Symbol: FDFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709437.3 | XP_002709483.1 | 2 - 3 | 351 | 67 | |
| XM_008248858.2 | XP_008247080.1 | 1 - 2 | 474 | 67 | ||
More Information
Set Membership: |
|

FDFT1
Genomic Map