Assay Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721908.3 | XP_002721954.1 | 1690 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF543| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721908.3 | XP_002721954.1 | 1690 | 86 | ||
Set Membership: |
|