Assay Details
| Gene Symbol: PDCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710014.3 | XP_002710060.1 | 3 - 4 | 338 | 68 | |
| XM_017339560.1 | XP_017195049.1 | 3 - 4 | 338 | 68 | ||
More Information
Set Membership: |
|

PDCL3
Genomic Map