Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346890.1 | XP_017202379.1 | 2 - 3 | 1092 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017346890.1 transcripts.
Set Membership: |
|

SPTSSB
Genomic Map