Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262642.1 | XP_008260864.1 | 2 - 3 | 560 | 64 | |
| XM_008262643.2 | XP_008260865.1 | 2 - 3 | 1623 | 64 | ||
More Information
Set Membership: |
|

ZFP57
Genomic Map