Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262318.2 | XP_008260540.1 | 5 - 6 | 1211 | 65 | |
| XM_008262319.2 | XP_008260541.1 | 5 - 6 | 1187 | 65 | ||
| XM_017344772.1 | XP_017200261.1 | 4 - 5 | 1026 | 65 | ||
More Information
Set Membership: |
|

OCLN
Genomic Map