Assay Details
| Gene Symbol: ADGRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714479.3 | XP_002714525.1 | 4 - 5 | 569 | 64 | |
| XM_008263022.2 | XP_008261244.1 | 5 - 6 | 700 | 64 | ||
| XM_008263023.2 | XP_008261245.1 | 6 - 7 | 810 | 64 | ||
| XM_017345169.1 | XP_017200658.1 | 5 - 6 | 679 | 64 | ||
More Information
Set Membership: |
|

ADGRF4
Genomic Map