Assay Details
| Gene Symbol: JMJD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338913.1 | XP_017194402.1 | 690 | 56 | ||
| XM_017338914.1 | XP_017194403.1 | 690 | 56 | |||
| XM_017338915.1 | XP_017194404.1 | 690 | 56 | |||
More Information
Set Membership: |
|

JMJD6
Genomic Map