Assay Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708183.3 | XP_002708229.1 | 17 - 18 | 1936 | 66 | |
| XM_008255921.2 | XP_008254143.1 | 17 - 18 | 1914 | 66 | ||
More Information
Set Membership: |
|

PUM3
Genomic Map