Assay Details
| Gene Symbol: GORASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260281.2 | XP_008258503.1 | 2 - 3 | 241 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GORASP1| Gene Symbol: GORASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260281.2 | XP_008258503.1 | 2 - 3 | 241 | 71 | |
Set Membership: |
|