Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710188.3 | XP_002710234.1 | 7 - 8 | 1121 | 74 | |
| XM_008255336.2 | XP_008253558.1 | 7 - 8 | 1088 | 74 | ||
More Information
Set Membership: |
|

RNF145
Genomic Map