Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269126.2 | XP_008267348.1 | 4 - 5 | 728 | 73 | |
| XM_008269127.1 | XP_008267349.1 | 4 - 5 | 653 | 73 | ||
More Information
Set Membership: |
|

LRRC57
Genomic Map