Assay Details
| Gene Symbol: WNT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712775.3 | XP_002712821.1 | 4 - 5 | 788 | 62 | |
| XM_008259814.2 | XP_008258036.1 | 4 - 5 | 825 | 62 | ||
| XM_017343437.1 | XP_017198926.1 | 4 - 5 | 745 | 62 | ||
More Information
Set Membership: |
|

WNT5B
Genomic Map