Assay Details
| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711000.3 | XP_002711046.1 | 5 - 6 | 633 | 63 | |
| XM_017341826.1 | XP_017197315.1 | 5 - 6 | 638 | 63 | ||
More Information
Set Membership: |
|

CCNT1
Genomic Map