Assay Details
| Gene Symbol: LOC100352045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721270.3 | XP_002721316.1 | 1042 | 62 | ||
| XM_017350804.1 | XP_017206293.1 | 574 | 62 | |||
| XM_017350805.1 | XP_017206294.1 | 591 | 62 | |||
More Information
Set Membership: |
|

LOC100352045
Genomic Map