Assay Details
| Gene Symbol: PAK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348147.1 | XP_017203636.1 | 7 - 8 | 1907 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAK6| Gene Symbol: PAK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348147.1 | XP_017203636.1 | 7 - 8 | 1907 | 56 | |
Set Membership: |
|