Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713622.3 | XP_002713668.1 | 13 - 14 | 2128 | 66 | |
| XM_002713623.3 | XP_002713669.1 | 13 - 14 | 2128 | 66 | ||
| XM_002713624.3 | XP_002713670.1 | 12 - 13 | 2056 | 66 | ||
| XM_008261433.2 | XP_008259655.1 | 13 - 14 | 2128 | 66 | ||
| XM_017344220.1 | XP_017199709.1 | 13 - 14 | 2128 | 66 | ||
| XM_017344221.1 | XP_017199710.1 | 13 - 14 | 2128 | 66 | ||
More Information
Set Membership: |
|

RELCH
Genomic Map