Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723738.3 | XP_002723784.2 | 111 | 69 | ||
| XM_008252415.2 | XP_008250637.2 | 111 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM143| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723738.3 | XP_002723784.2 | 111 | 69 | ||
| XM_008252415.2 | XP_008250637.2 | 111 | 69 | |||
Set Membership: |
|