Assay Details
| Gene Symbol: ZNF268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250009.2 | XP_008248231.2 | 1187 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF268| Gene Symbol: ZNF268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250009.2 | XP_008248231.2 | 1187 | 78 | ||
Set Membership: |
|