Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268862.2 | XP_008267084.1 | 2 - 3 | 246 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAPC5| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268862.2 | XP_008267084.1 | 2 - 3 | 246 | 72 | |
Set Membership: |
|