Assay Details
| Gene Symbol: PPIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714673.3 | XP_002714719.1 | 3 - 4 | 1129 | 85 | |
| XM_017345079.1 | XP_017200568.1 | 3 - 4 | 983 | 85 | ||
More Information
Set Membership: |
|

PPIL1
Genomic Map