Assay Details
| Gene Symbol: ESRRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717508.3 | XP_002717554.1 | 2 - 3 | 630 | 62 | |
| XM_008268418.2 | XP_008266640.1 | 3 - 4 | 709 | 62 | ||
| XM_008268419.2 | XP_008266641.1 | 3 - 4 | 581 | 62 | ||
| XM_008268424.2 | XP_008266646.1 | 2 - 3 | 590 | 62 | ||
More Information
Set Membership: |
|

ESRRG
Genomic Map