Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272226.2 | XP_008270448.1 | 4 - 5 | 421 | 66 | |
| XM_008272227.2 | XP_008270449.1 | 5 - 6 | 460 | 66 | ||
| XM_008272231.2 | XP_008270453.1 | 3 - 4 | 381 | 66 | ||
| XM_017349524.1 | XP_017205013.1 | 4 - 5 | 422 | 66 | ||
More Information
Set Membership: |
|

HVCN1
Genomic Map