Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256196.2 | XP_008254418.1 | 3 - 4 | 1789 | 88 | |
| XM_008256198.2 | XP_008254420.1 | 3 - 4 | 1790 | 88 | ||
| XM_017341722.1 | XP_017197211.1 | 3 - 4 | 1740 | 88 | ||
More Information
Set Membership: |
|

SIRPB2
Genomic Map