Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256196.2 | XP_008254418.1 | 5 - 6 | 2023 | 77 | |
| XM_008256197.2 | XP_008254419.1 | 5 - 6 | 2022 | 77 | ||
| XM_017341722.1 | XP_017197211.1 | 5 - 6 | 1974 | 77 | ||
More Information
Set Membership: |
|

SIRPB2
Genomic Map