Assay Details
| Gene Symbol: FOXK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251176.2 | XP_008249398.2 | 1297 | 67 | ||
| XM_017339051.1 | XP_017194540.1 | 1297 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXK2| Gene Symbol: FOXK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008251176.2 | XP_008249398.2 | 1297 | 67 | ||
| XM_017339051.1 | XP_017194540.1 | 1297 | 67 | |||
Set Membership: |
|