Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715405.3 | XP_002715451.1 | 1 - 2 | 170 | 68 | |
| XM_008264606.2 | XP_008262828.1 | 1 - 2 | 177 | 68 | ||
More Information
Set Membership: |
|

CD2
Genomic Map