Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720186.3 | XP_002720232.1 | 4 - 5 | 764 | 96 | |
| XM_008272869.2 | XP_008271091.1 | 3 - 4 | 549 | 96 | ||
More Information
Set Membership: |
|

NXT2
Genomic Map