Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269748.2 | XP_008267970.1 | 2 - 3 | 430 | 59 | |
| XM_017348348.1 | XP_017203837.1 | 2 - 3 | 430 | 59 | ||
| XM_017348349.1 | XP_017203838.1 | 2 - 3 | 431 | 59 | ||
More Information
Set Membership: |
|

OCA2
Genomic Map