Assay Details
Gene Symbol: LYRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711812.3 | XP_002711858.1 | 3 - 4 | 517 | 79 | |
XM_008257832.2 | XP_008256054.1 | 4 - 5 | 746 | 79 | ||
XM_008257833.2 | XP_008256055.1 | 4 - 5 | 498 | 79 | ||
XM_008257835.2 | XP_008256057.1 | 5 - 6 | 550 | 79 | ||
XM_008257836.2 | XP_008256058.1 | 4 - 5 | 394 | 79 | ||
XM_017342506.1 | XP_017197995.1 | 3 - 4 | 277 | 79 | ||
XM_017342507.1 | XP_017197996.1 | 5 - 6 | 575 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
