Assay Details
| Gene Symbol: AAGAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718171.3 | XP_002718217.1 | 9 - 10 | 935 | 64 | |
| XM_008268855.2 | XP_008267077.1 | 9 - 10 | 938 | 64 | ||
More Information
Set Membership: |
|

AAGAB
Genomic Map