Assay Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270779.2 | XP_008269001.2 | 2 - 3 | 216 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFSF12| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270779.2 | XP_008269001.2 | 2 - 3 | 216 | 76 | |
Set Membership: |
|